• USA Home
  • 37745 - AQUANAL-professional ortho-Phosphate tube test
EMAIL THIS PAGE TO A FRIEND
37745 Sigma-Aldrich
AQUANAL-professional ortho-Phosphate tube test
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
measuring range   3.0-60 mg/L
Description
Analysis Note
Method: forming of a yellow colored molybdatophosphorous complex
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
24 reaction tubes, 1 blank tube
General description
Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations. AQUANAL-professional tube tests were developed to provide a fast safe and clean method in order to perform water analysis in sewage treatment plants. The sample is pipetted into a tube that contains the test reagent. For some tests, an additional reagent is required that has to be added manually.
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2796 8 / PGII
WGK Germany 
1
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?