• USA Home
  • 37761 - 2,3-Dihydroxynaphthalene

EMAIL THIS PAGE TO A FRIEND
37761 Sigma-Aldrich

2,3-Dihydroxynaphthalene

technical, ≥90% (HPLC)

Synonym: 2,3-Naphthalenediol

  • CAS Number 92-44-4

  • Linear Formula C10H6(OH)2

  • Molecular Weight 160.17

  •  Beilstein Registry Number 742375

  •  EC Number 202-156-7

  •  MDL number MFCD00004073

  •  PubChem Substance ID 329755692

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
InChI Key   JRNGUTKWMSBIBF-UHFFFAOYSA-N
grade   technical
assay   ≥90% (HPLC)
mp   161-165 °C(lit.)
  161-165 °C
Gene Information   human ... BAD(572)

Description

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
QJ4750000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?