• USA Home
  • 37765 - 2,6-Dihydroxynaphthalene

EMAIL THIS PAGE TO A FRIEND
37765 Sigma-Aldrich

2,6-Dihydroxynaphthalene

purum, ≥96.0% (HPLC)

Synonym: 2,6-Naphthalenediol

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
InChI Key   MNZMMCVIXORAQL-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (HPLC)
mp   220-224 °C
  223-225 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

275603 2,6-Dihydroxynaphthalene, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?