• USA Home
  • 37791 - AQUANAL-plus carbonate hardness

EMAIL THIS PAGE TO A FRIEND
37791 Sigma-Aldrich

AQUANAL-plus carbonate hardness

check solution for 37437 (5°KH)

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
grade   check solution for 37437 (5°KH)

Description

General description

Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations. Carbonate Hardness is part of total hardness. The calcium and magnesium ions, which are present as carbonate and hydrogen carbonate are referred to as carbonate hardness or temporary hardness. The carbonate hardness is determined by titration with hydrochloric acid. The end point of titration is indicated by the Cooper′s indicator. Carbonate hardness is a measure of the sensitivity of the water to pH variations and is especially important in aquariums and water supply companies.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?