EMAIL THIS PAGE TO A FRIEND
37960 Sigma-Aldrich

2-Methylresorcinol

technical, ≥90% (GC)

Synonym: 2,6-Dihydroxytoluene

  • CAS Number 608-25-3

  • Linear Formula CH3C6H3(OH)2

  • Molecular Weight 124.14

  •  Beilstein Registry Number 2042177

  •  EC Number 210-155-8

  •  MDL number MFCD00002271

  •  PubChem Substance ID 24863673

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
InChI Key   ZTMADXFOCUXMJE-UHFFFAOYSA-N
grade   technical
assay   ≥90% (GC)
bp   264 °C(lit.)
mp   114-120 °C(lit.)
  114-121 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
VH2009500
Flash Point(F) 
275 °F
Flash Point(C) 
135 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?