• USA Home
  • 38100 - 3,5-Diiodo-L-thyronine

EMAIL THIS PAGE TO A FRIEND
38100 Sigma-Aldrich

3,5-Diiodo-L-thyronine

≥96.0% (calc. based on dry substance, NT)

Synonym: 3,5-Diiodo-4-(4-hydroxyphenoxy)-L-phenylalanine, 3,5-T2, O-(4-Hydroxyphenyl)-3,5-diiodo-L-tyrosine

  • CAS Number 1041-01-6

  • Empirical Formula (Hill Notation) C15H13I2NO4

  • Molecular Weight 525.08

  •  Beilstein Registry Number 2707891

  •  EC Number 213-867-7

  •  MDL number MFCD00064987

Purchase

Properties

InChI Key   ZHSOTLOTTDYIIK-ZDUSSCGKSA-N
assay   ≥96.0% (calc. based on dry substance, NT)
optical activity   [α]20/D +28±2°, c = 1% in ethanol/1 M HCl (2:1)
impurities   ≤5% water
mp   255-260 °C (dec.)
storage temp.   2-8°C

Description

Other Notes

Preparation of high specific activity labeled triiodothyronine for radioimmunoassay

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

D0629 3,5-Diiodo-L-thyronine, thyroid hormone analog

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?