• USA Home
  • 38125 - Vinyl chloride in nitrogen

EMAIL THIS PAGE TO A FRIEND
38125 Sigma-Aldrich

Vinyl chloride in nitrogen

analytical standard, 10 ppm

Synonym: Chloroethylene in nitrogen

Purchase

Properties

grade   analytical standard
form   gas
shelf life   limited shelf life, expiry date on the label
concentration   10 ppm
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   multi-component solution

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS04  GHS04
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1956 2.2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?