EMAIL THIS PAGE TO A FRIEND
38300 Sigma-Aldrich

Diisopropylamine

purum, ≥98.0% (GC)

Synonym: DIPA

  • CAS Number 108-18-9

  • Linear Formula (CH3)2CHNHCH(CH3)2

  • Molecular Weight 101.19

  •  Beilstein Registry Number 605284

  •  EC Number 203-558-5

  •  MDL number MFCD00008862

Purchase

Properties

vapor density   3.5 (vs air)
vapor pressure   50 mmHg ( 20 °C)
InChI Key   UAOMVDZJSHZZME-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
autoignition temp.   599 °F
expl. lim.   8.5 %
refractive index   n20/D 1.392(lit.)
  n20/D 1.393
bp   84 °C(lit.)
mp   −61 °C(lit.)
density   0.722 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1158 - class 3 - PG 2 - Diisopropylamine
WGK Germany 
2
RTECS 
IM4025000
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Related Products

replaced by

Product #

Description

Add to Cart

471224 Diisopropylamine, ≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?