CAS Number 5970-45-6
Linear Formula Zn(CH3COO)2 · 2H2O
Molecular Weight 219.51
Beilstein Registry Number 3732513
EC Number 209-170-2
MDL number MFCD00066961
PubChem Substance ID 24863930
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| assay | ≥98% |
| 98.0-101.0% (ACS specification) | |
| impurities | ≤0.005% insolubles |
| pH | 6.0-7.0 (25 °C, 5%) |
| anion traces | chloride (Cl-): ≤5 ppm |
| sulfate (SO42-): ≤0.005% |
12 kg in poly drum
500 g in poly bottle
2.5 kg in poly bottle
Employed in the synthesis of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis.1 Also used in the ultrasonic preparation of zinc sulfide nanoparticles coated on silica particles.2
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?