• USA Home
  • 38345 - 2,6-Diisopropylaniline

EMAIL THIS PAGE TO A FRIEND
38345 Sigma-Aldrich

2,6-Diisopropylaniline

technical, ~90% (GC)

Purchase

Properties

vapor pressure   <0.01 mmHg ( 20 °C)
InChI Key   WKBALTUBRZPIPZ-UHFFFAOYSA-N
grade   technical
assay   ~90% (GC)
refractive index   n20/D 1.532(lit.)
  n20/D 1.532
bp   257 °C(lit.)
mp   −45 °C(lit.)
density   0.94 g/mL at 25 °C(lit.)

Description

Application

2,6-Diisopropylaniline may be used in the preparation of multitopic Schiff-base ligand precursors. It may be used in preparation of NSN-donor proligand, 4,5-bis(2,6-diisopropylanilino)-2,7-di-tert-butyl-9,9-dimethylthioxanthene.

General description

2,6-Diisopropylaniline on condensation with triacetylmethane in toluene in the presence of p-toluenesulfonic acid provides 3-[1-(2,6-diisopropylphenylamino)ethylidene]pentane-2,4-dione.

Price and Availability

Safety & Documentation

Safety Information

Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
BX4025000
Flash Point(F) 
242.6 °F
Flash Point(C) 
117 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?