• USA Home
  • 38353 - 1,4-Diisopropylbenzene

EMAIL THIS PAGE TO A FRIEND
38353 Sigma-Aldrich

1,4-Diisopropylbenzene

analytical standard

  • CAS Number 100-18-5

  • Linear Formula C6H4[CH(CH3)2]2

  • Molecular Weight 162.27

  •  Beilstein Registry Number 1854739

  •  EC Number 202-826-9

  •  MDL number MFCD00008892

  •  PubChem Substance ID 24863975

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard
vapor density   5.6 (vs air)
vapor pressure   0.25 mmHg ( 20 °C)
InChI Key   SPPWGCYEYAMHDT-UHFFFAOYSA-N
assay   ≥99.5% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.489(lit.)
  n20/D 1.490
bp   210 °C(lit.)
mp   −17 °C(lit.)
density   0.857 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
RTECS 
CZ6360000
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?