• USA Home
  • 38433 - Diisopropoxytitanium bis(acetylacetonate) solution

EMAIL THIS PAGE TO A FRIEND
38433 Sigma-Aldrich

Diisopropoxytitanium bis(acetylacetonate) solution

purum, ~75% in isopropanol

Synonym: Ti(acac)2OiPr2

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Titanium More...
InChI Key   OVSGBKZKXUMMHS-VVDZMTNVSA-L
grade   purum
concentration   ~75% in isopropanol
density   1.00 g/mL at 20 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1219 - class 3 - PG 2 - Isopropanol, solution
WGK Germany 
3
Flash Point(F) 
53.6 °F
Flash Point(C) 
12 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?