• USA Home
  • 38530 - Sodium 2,3-dimercaptopropanesulfonate monohydrate

EMAIL THIS PAGE TO A FRIEND
38530 Sigma-Aldrich

Sodium 2,3-dimercaptopropanesulfonate monohydrate

purum, ≥97.0% (T)

Synonym: 2,3-Dimercaptopropanesulfonic acid sodium salt monohydrate, DMPS

  • CAS Number 207233-91-8

  • Linear Formula HSCH2CH(SH)CH2SO3Na · H2O

  • Molecular Weight 228.29

  •  Beilstein Registry Number 3734863

  •  EC Number 223-796-3

  •  MDL number MFCD00149543

  •  PubChem Substance ID 329755770

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acids, Sulfur Compounds More...
InChI Key   XMUHNMQFDVIWGU-UHFFFAOYSA-M
grade   purum
assay   ≥97.0% (T)
mp   215 °C (dec.)
solubility   H2O: soluble0.1 g/mL, clear

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D8016 Sodium 2,3-dimercaptopropanesulfonate monohydrate, ~95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?