• USA Home
  • 38558 - 3′,4′-Dimethoxyacetophenone

EMAIL THIS PAGE TO A FRIEND
38558 Sigma-Aldrich

3′,4′-Dimethoxyacetophenone

purum, ≥97.0% (GC)

  • CAS Number 1131-62-0

  • Linear Formula (CH3O)2C6H3COCH3

  • Molecular Weight 180.20

  •  Beilstein Registry Number 781213

  •  EC Number 214-468-0

  •  MDL number MFCD00008737

  •  PubChem Substance ID 24864039

Purchase

Properties

Related Categories Building Blocks, C10, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   IQZLUWLMQNGTIW-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
bp   286-288 °C(lit.)
mp   47-51 °C
  47-54 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?