• USA Home
  • 386189 - 1-(3-Sulfopropyl)-2-(2-{[1-(3-sulfopropyl)naphtho[1,2-d]thiazol-2(1H)-ylidene]methyl}-1-butenyl)naphtho[1,2-d]thiazoliumhydroxide inner salt, triethylammonium salt

EMAIL THIS PAGE TO A FRIEND
386189 Sigma-Aldrich

1-(3-Sulfopropyl)-2-(2-{[1-(3-sulfopropyl)naphtho[1,2-d]thiazol-2(1H)-ylidene]methyl}-1-butenyl)naphtho[1,2-d]thiazoliumhydroxide inner salt, triethylammonium salt

Dye content 95 %

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, S, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   IRGJPQDDBXQCAP-UHFFFAOYSA-N
composition   Dye content, 95%
mp   230 °C (dec.)(lit.)
absorption   λmax 579 nm

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?