EMAIL THIS PAGE TO A FRIEND
38936 Sigma-Aldrich

Dimethylamine

purum, ≥99.0%

Purchase

Properties

Related Categories Chemical Synthesis, Specialty Gases, Synthetic Reagents More...
vapor density   1.55 (15 °C, vs air)
vapor pressure   1277 mmHg ( 20 °C)
InChI Key   ROSDSFDQCJNGOL-UHFFFAOYSA-N
grade   purum
assay   ≥99.0%
autoignition temp.   753 °F
expl. lim.   14.4 %
impurities   ≤0.05% ammonia
  ≤0.2% methylamine
  ≤0.2% trimethylamine
bp   7 °C(lit.)
mp   −93 °C(lit.)
density   0.68 g/mL at 20 °C(lit.)

Description

Other Notes

Sales restrictions may apply

Packaging

Cylinder with net ~15 kg

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1032 - class 2 - Dimethylamine, anhydrous
WGK Germany 
2
RTECS 
IP8750000
Flash Point(F) 
21.2 °F
Flash Point(C) 
-6 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

control valve

Product #

Description

Add to Cart

99112 Outlet valve No 70688, stainless steel, pkg of 1 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?