• USA Home
  • 39147 - D-(+)-α-Dimethylamino-ε-caprolactam

EMAIL THIS PAGE TO A FRIEND
39147 Sigma-Aldrich

D-(+)-α-Dimethylamino-ε-caprolactam

puriss., ≥99.0% (NT)

Synonym: (R)-3-Dimethylamino-hexahydro-2-azepinone

  • CAS Number 88764-67-4

  • Empirical Formula (Hill Notation) C8H16N2O

  • Molecular Weight 156.23

  •  MDL number MFCD00066074

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Lactams, Organic Building Blocks More...
InChI Key   CFZGIDYCUWFUJR-SSDOTTSWSA-N
grade   puriss.
assay   ≥99.0% (NT)
optical activity   [α]20/D +27.5±1°, c = 3% in H2O
mp   58-60 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
330.8 °F
Flash Point(C) 
166 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?