• USA Home
  • 39416 - 2-[4-(Dimethylamino)styryl]-1-methylquinolinium iodide

EMAIL THIS PAGE TO A FRIEND
39416 Sigma-Aldrich

2-[4-(Dimethylamino)styryl]-1-methylquinolinium iodide

for fluorescence, ≥98.0% (TLC)

  • CAS Number 3915-61-5

  • Empirical Formula (Hill Notation) C20H21IN2

  • Molecular Weight 416.30

  •  Beilstein Registry Number 6102208

  •  MDL number MFCD00031815

  •  PubChem Substance ID 329755868

Purchase

Properties

Related Categories Biochemicals and Reagents, Detection, Fluorescent Probes, Labels, Particles and Stains, Fluorescent Stains, Nucleic Acid Stains,
grade   for fluorescence
InChI Key   VGLNXCCOYCGRLG-UHFFFAOYSA-M
assay   ≥98.0% (TLC)
mp   261 °C (dec.)(lit.)
solubility   DMSO: soluble
  methanol: soluble
fluorescence   λex 463 nm; λem 512 nm in phosphate buffer/SDS pH 7.0

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?