EMAIL THIS PAGE TO A FRIEND
39550 Sigma-Aldrich

3,5-Dimethylaniline

technical, ≥97.0% (GC)

Synonym: 1-Amino-3,5-dimethylbenzene, 3,5-Xylidine, 5-Amino-m-xylene

  • CAS Number 108-69-0

  • Linear Formula (CH3)2C6H3NH2

  • Molecular Weight 121.18

  •  Beilstein Registry Number 507051

  •  EC Number 203-607-0

  •  MDL number MFCD00007813

  •  PubChem Substance ID 329755879

Purchase

Properties

InChI Key   MKARNSWMMBGSHX-UHFFFAOYSA-N
grade   technical
assay   ≥97.0% (GC)
refractive index   n20/D 1.557(lit.)
  n20/D 1.557
bp   104-105 °C/14 mmHg(lit.)
mp   7-9 °C
density   0.972 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1711 6.1 / PGII
WGK Germany 
3
Flash Point(F) 
213.8 °F
Flash Point(C) 
101 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?