• USA Home
  • 39581 - 2,3-Dimethylbenzoic acid

EMAIL THIS PAGE TO A FRIEND
39581 Sigma-Aldrich

2,3-Dimethylbenzoic acid

purum, ≥97.0% (T)

Synonym: vic.-o-Xylylic acid

  • CAS Number 603-79-2

  • Linear Formula (CH3)2C6H3CO2H

  • Molecular Weight 150.17

  •  Beilstein Registry Number 971590

  •  EC Number 210-058-0

  •  MDL number MFCD00002479

Purchase

Properties

InChI Key   RIZUCYSQUWMQLX-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (T)
ign. residue   ~0.5%
mp   144-146 °C(lit.)
  144-146 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DG8734000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

138150 2,3-Dimethylbenzoic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?