• USA Home
  • 39582 - 2,4-Dimethylbenzoic acid

EMAIL THIS PAGE TO A FRIEND
39582 Sigma-Aldrich

2,4-Dimethylbenzoic acid

purum, ≥98.0% (T)

Synonym: asym.-m-Xylylic acid

  • CAS Number 611-01-8

  • Linear Formula (CH3)2C6H3CO2H

  • Molecular Weight 150.17

  •  Beilstein Registry Number 971589

  •  EC Number 210-246-2

  •  MDL number MFCD00002480

  •  PubChem Substance ID 24864172

Purchase

Properties

Related Categories Building Blocks, C9, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   BKYWPNROPGQIFZ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
ign. residue   ≤0.3%
bp   267 °C/727 mmHg(lit.)
mp   124-126 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?