• USA Home
  • 39600 - 3,5-Dimethylbenzoic acid

EMAIL THIS PAGE TO A FRIEND
39600 Sigma-Aldrich

3,5-Dimethylbenzoic acid

purum, ≥97.0% (HPLC)

Synonym: Mesitylenic acid, sym.-m-Xylylic acid

  • CAS Number 499-06-9

  • Linear Formula (CH3)2C6H3CO2H

  • Molecular Weight 150.17

  •  Beilstein Registry Number 1072182

  •  EC Number 207-876-5

  •  MDL number MFCD00002525

  •  PubChem Substance ID 329755886

Purchase

Properties

Related Categories Building Blocks, C9, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   UMVOQQDNEYOJOK-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (HPLC)
ign. residue   ≤0.1%
mp   169-171 °C(lit.)
  169-171 °C
solubility   methanol: soluble1 g/10 mL, clear, colorless

Description

Application

3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp. WR912.

General description

3,5-Dimethylbenzoic acid forms inclusion complex with β-cyclodextrin and its crystal structure is reported to be triclinic.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DG8734030

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?