Synonym: Ammonium vanadate(V)
CAS Number 7803-55-6
Linear Formula NH4VO3
Molecular Weight 116.98
EC Number 232-261-3
MDL number MFCD00011430
PubChem Substance ID 24864719
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| assay | ≥99.0% |
| solubility | NH4OH: passes test |
| density | 2.32 g/cm3 at 25 °C(lit.) |
| anion traces | carbonate (CO32-): passes test |
| chloride (Cl-): ≤0.2% | |
| sulfate (SO42-): ≤0.05% |
50, 250 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?