• USA Home
  • 39860 - Trimethylsilyl N,N-dimethylcarbamate

EMAIL THIS PAGE TO A FRIEND
39860 Sigma-Aldrich

Trimethylsilyl N,N-dimethylcarbamate

purum, ≥95.0% (GC)

Synonym: DMCTMS

  • CAS Number 32115-55-2

  • Linear Formula (CH3)2NCO2Si(CH3)3

  • Molecular Weight 161.27

  •  Beilstein Registry Number 1928814

  •  MDL number MFCD00042933

  •  PubChem Substance ID 329755912

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Protected Amines More...
InChI Key   RDEKHAYBXPYQJQ-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.418(lit.)
  n20/D 1.418
bp   74-75 °C/20 mmHg(lit.)
density   0.925 g/mL at 25 °C(lit.)

Description

Other Notes

Silylating agent for alcohols, phenols and carboxylic acids

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
55.4 °F
Flash Point(C) 
13 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?