• USA Home
  • 39871 - Dimethylcarbamoyl chloride

EMAIL THIS PAGE TO A FRIEND
39871 Sigma-Aldrich

Dimethylcarbamoyl chloride

purum, ≥98.0% (AT)

Synonym: Chloroformic acid dimethyl amide

  • CAS Number 79-44-7

  • Linear Formula (CH3)2NCOCl

  • Molecular Weight 107.54

  •  Beilstein Registry Number 878197

  •  EC Number 201-208-6

  •  MDL number MFCD00000635

  •  PubChem Substance ID 329755915

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
InChI Key   YIIMEMSDCNDGTB-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)

Description

Other Notes

This product has been replaced by D152803-ALDRICH | Dimethylcarbamyl chloride 98%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2262 8 / PGII
WGK Germany 
3
Flash Point(F) 
179.6 °F
Flash Point(C) 
82 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D152803 Dimethylcarbamyl chloride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?