• USA Home
  • 40160 - Dimethoxydimethylsilane

EMAIL THIS PAGE TO A FRIEND
40160 Sigma-Aldrich

Dimethoxydimethylsilane

purum, ≥96.0% (GC)

Synonym: Dimethyldimethoxysilane

  • CAS Number 1112-39-6

  • Linear Formula Si(OCH3)2(CH3)2

  • Molecular Weight 120.22

  •  Beilstein Registry Number 1733694

  •  EC Number 214-189-4

  •  MDL number MFCD00025691

  •  PubChem Substance ID 24865029

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organosilicon, Other Organosilicon
InChI Key   JJQZDUKDJDQPMQ-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
form   liquid
refractive index   n20/D 1.369(lit.)
  n20/D 1.371
bp   81.4 °C(lit.)
density   0.88 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
3
RTECS 
VV3641666
Flash Point(F) 
50 °F
Flash Point(C) 
10 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?