• USA Home
  • 40246 - N,N-Dimethylformamide

EMAIL THIS PAGE TO A FRIEND
40246 Sigma-Aldrich

N,N-Dimethylformamide

B&J Brand (product of Burdick & Jackson)

Synonym: DMF, NSC 5356

  • CAS Number 68-12-2

  • Linear Formula HCON(CH3)2

  • Molecular Weight 73.09

  •  Beilstein Registry Number 605365

  •  EC Number 200-679-5

  •  MDL number MFCD00003284

  •  PubChem Substance ID 329755959

Purchase

Properties

Related Categories Dimethylformamide (DMF), Solvent by Type, Solvents More...
grade   B&J Brand (product of Burdick & Jackson)
vapor density   2.5 (vs air)
vapor pressure   2.7 mmHg ( 20 °C)
InChI Key   ZMXDDKWLCZADIW-UHFFFAOYSA-N
autoignition temp.   833 °F
expl. lim.   15.2 %
refractive index   n20/D 1.430(lit.)
bp   153 °C(lit.)
mp   −61 °C(lit.)
density   0.944 g/mL(lit.)

Description

Application

Solvent for many hydrophobic organic compounds.

Other Notes

Sales restrictions may apply

General description

Special brochure (85475) available at cost price.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2265 3 / PGIII
WGK Germany 
2
RTECS 
LQ2100000
Flash Point(F) 
136.4 °F
Flash Point(C) 
58 °C
Protocols & Articles

Related Content

Properties of Solvents Table

a. Gant, R., LC/GC 10(7), 515 (1992) 1. At 20 °C 2. At 20 °C +/- 5 °C relative to water at 4 °C 3. At 10mmHg pressure 4. At l=589.32nm
Keywords: Gas chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?