402893 Sigma-Aldrich

1-Propanol

ACS reagent, ≥99.5%

DOWNLOAD MSDS (PDF)

Synonym: Propyl alcohol

Properties

Related Categories ACS Grade, ACS Grade Solvents, ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Flex-Spout Cans,
grade   ACS reagent
vapor density   2.1 (vs air)
vapor pressure   10 mmHg ( 147 °C)
  14.9 mmHg ( 20 °C)
assay   ≥99.5%
autoignition temp.   700 °F
expl. lim.   13.7 %
impurities   ≤0.0004 meq/g Titr. acid
  ≤0.01% ethanol
  ≤0.01% methanol
  ≤0.03% Carbonyl compounds (as C2H5CHO)
  ≤0.05% CH3CHOHCH3
  ≤0.05% isopropanol
  ≤0.2% water
evapn. residue   ≤0.001%
color   APHA: ≤10
refractive index   n20/D 1.384(lit.)
bp   97 °C(lit.)
mp   −127 °C(lit.)
solubility   H2O: passes test
density   0.804 g/mL at 25 °C(lit.)

Description

Packaging

1, 2.5, 4×4 L in glass bottle

20 L in steel drum

500 mL in glass bottle

Price and Availability

Customers Also Viewed

1-Butanol
3604651-Butanol

ACS reagent, ≥99.4%

1-Propanol

anhydrous, 99.7%

1-Propanol

CHROMASOLV®, for HPLC, ≥99.9%

1-Propanol

analytical standard

2-Propanol

anhydrous, 99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?