Synonym: Glauber’s salt
CAS Number 7727-73-3
Linear Formula Na2SO4 · 10H2O
Molecular Weight 322.20
EC Number 231-820-9
MDL number MFCD00149189
PubChem Substance ID 24865152
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Chemical Synthesis, Essential Chemicals, Inorganic Salts, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| assay | ≥99.0% |
| impurities | ≤0.01% insolubles |
| pH | 5.2-9.2 (25 °C, 5%) |
| mp | 32.4 °C |
| density | 1.46 g/mL at 25 °C(lit.) |
| anion traces | chloride (Cl-): ≤5 ppm |
100, 500 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?