EMAIL THIS PAGE TO A FRIEND
40360 Sigma-Aldrich

N,N-Dimethylglycine

puriss., ≥99.0% (NT)

Synonym: (Dimethylamino)acetic acid

  • CAS Number 1118-68-9

  • Linear Formula (CH3)2NCH2COOH

  • Molecular Weight 103.12

  •  Beilstein Registry Number 1700261

  •  EC Number 214-267-8

  •  MDL number MFCD00004283

  •  PubChem Substance ID 24865204

Purchase

Properties

Related Categories Amino Acids, Core Bioreagents, Research Essentials More...
InChI Key   FFDGPVCHZBVARC-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (NT)
mp   178-182 °C(lit.)
  178-182 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
MB9865000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

D1156 N,N-Dimethylglycine, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?