• USA Home
  • 40520 - 2,5-Dimethyl-2,5-hexanediol

EMAIL THIS PAGE TO A FRIEND
40520 Sigma-Aldrich

2,5-Dimethyl-2,5-hexanediol

purum, ≥97.0% (GC)

  • CAS Number 110-03-2

  • Linear Formula (CH3)2C(OH)CH2CH2C(OH)(CH3)2

  • Molecular Weight 146.23

  •  Beilstein Registry Number 1361437

  •  EC Number 203-731-5

  •  MDL number MFCD00004473

  •  PubChem Substance ID 329755998

Purchase

Properties

InChI Key   ZWNMRZQYWRLGMM-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
bp   214-215 °C(lit.)
mp   86-90 °C(lit.)
  87-89 °C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
260 °F
Flash Point(C) 
127 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

143618 2,5-Dimethyl-2,5-hexanediol, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?