• USA Home
  • 40600 - (R)-2-Piperazinecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
40600 Sigma-Aldrich

(R)-2-Piperazinecarboxylic acid

  • CAS Number 31321-68-3

  • Empirical Formula (Hill Notation) C5H10N2O2

  • Molecular Weight 130.15

Purchase

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?