• USA Home
  • 40660 - N,N-Dimethylhexylamine

EMAIL THIS PAGE TO A FRIEND
40660 Sigma-Aldrich

N,N-Dimethylhexylamine

purum, ≥96.0% (GC)

  • CAS Number 4385-04-0

  • Linear Formula CH3(CH2)4CH2N(CH3)2

  • Molecular Weight 129.24

  •  Beilstein Registry Number 1732757

  •  EC Number 224-491-8

  •  MDL number MFCD00009522

  •  PubChem Substance ID 24865389

Purchase

Properties

InChI Key   QMHNQZGXPNCMCO-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
refractive index   n20/D 1.414(lit.)
  n20/D 1.414
bp   146-150 °C(lit.)
density   0.744 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2733PSN1 8(3) / PGIII
WGK Germany 
3
Flash Point(F) 
93.2 °F
Flash Point(C) 
34 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

308102 N,N-Dimethylhexylamine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?