CAS Number 12125-01-8
Linear Formula NH4F
Molecular Weight 37.04
MDL number MFCD00011423
PubChem Substance ID 24865437
Popular Documents: Specification Sheet (PDF)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?