• USA Home
  • 407674 - Silicon 2,3-naphthalocyanine dioctyloxide

EMAIL THIS PAGE TO A FRIEND
407674 Sigma-Aldrich

Silicon 2,3-naphthalocyanine dioctyloxide

Dye content 90 %

Synonym: 2,3-Naphthalocyaninato-dioctyloxysilane, Dioctyloxysilicon 2,3-naphthalocyanine

Purchase

Properties

Related Categories Infrared (IR) Dyes, Materials Science, Organic and Printed Electronics, Photonic and Optical Materials, Phthalocyanine and Porphyrin Dyes,
InChI Key   ZRBNRHRJAGPWCD-LLDQDFBRSA-N
composition   Dye content, 90%
absorption   λmax 798 nm

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?