CAS Number 83482-57-9
Linear Formula C6H5C(CH3)2C6H4OC6H3-1,2-(CN)2
Molecular Weight 338.40
MDL number MFCD00059048
PubChem Substance ID 24865599
| Related Categories | C, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes |
| assay | 97% |
| mp | 90-92 °C(lit.) |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?