• USA Home
  • 40930 - N,N-Dimethylnonylamine

EMAIL THIS PAGE TO A FRIEND
40930 Sigma-Aldrich

N,N-Dimethylnonylamine

purum, 98% (GC)

Synonym: 1-(Dimethylamino)nonane

  • CAS Number 17373-27-2

  • Linear Formula CH3(CH2)8N(CH3)2

  • Molecular Weight 171.32

  •  Beilstein Registry Number 1736553

  •  MDL number MFCD00053419

  •  PubChem Substance ID 329756036

Purchase

Properties

InChI Key   AMAADDMFZSZCNT-UHFFFAOYSA-N
grade   purum
assay   98% (GC)
refractive index   n20/D 1.428(lit.)
  n20/D 1.428
bp   83 °C/7 mmHg(lit.)
density   0.773 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?