• USA Home
  • 40958 - Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride

EMAIL THIS PAGE TO A FRIEND
40958 Sigma-Aldrich

Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride

technical, ~50% (AT)

Synonym: DMOAP

  • CAS Number 27668-52-6

  • Linear Formula [(CH3O)3Si(CH2)3N(CH3)2(CH2)17CH3]Cl

  • Molecular Weight 496.28

  •  Beilstein Registry Number 4897339

  •  MDL number MFCD00054224

  •  PubChem Substance ID 24865659

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   WSFMFXQNYPNYGG-UHFFFAOYSA-M
grade   technical
assay   ~50% (AT)
density   0.89 g/mL at 20 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1230 3/PG 2
WGK Germany 
3
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?