• USA Home
  • 41029 - cis-2,3-Dimethyloxirane

EMAIL THIS PAGE TO A FRIEND
41029 Sigma-Aldrich

cis-2,3-Dimethyloxirane

purum, ≥97.0% (GC)

Synonym: cis-2,3-Epoxybutane, cis-2-Butene oxide

  • CAS Number 1758-33-4

  • Empirical Formula (Hill Notation) C4H8O

  • Molecular Weight 72.11

  •  Beilstein Registry Number 79770

  •  MDL number MFCD00005128

  •  PubChem Substance ID 329756049

Purchase

Properties

InChI Key   PQXKWPLDPFFDJP-ZXZARUISSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.382
density   0.827 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3271 3/PG 2
WGK Germany 
3
RTECS 
EK3855020
Flash Point(F) 
-15 °F
Flash Point(C) 
-26 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

294047 cis-2,3-Epoxybutane, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?