• USA Home
  • 41385 - (R)-(+)-N,N,α-Trimethylbenzylamine

EMAIL THIS PAGE TO A FRIEND
41385 Sigma-Aldrich

(R)-(+)-N,N,α-Trimethylbenzylamine

purum, ≥95.0% (sum of enantiomers, GC)

Synonym: (R)-(+)-N,N-Dimethyl-1-phenylethylamine

  • CAS Number 19342-01-9

  • Empirical Formula (Hill Notation) C10H15N

  • Molecular Weight 149.23

  •  Beilstein Registry Number 3030178

  •  EC Number 242-976-2

  •  MDL number MFCD00008857

  •  PubChem Substance ID 329756096

Purchase

Properties

InChI Key   BVURNMLGDQYNAF-SECBINFHSA-N
grade   purum
assay   ≥95.0% (sum of enantiomers, GC)
optical activity   [α]20/D +49.5±2°, c = 1% in methanol
refractive index   n20/D 1.503(lit.)
bp   ~193 °C(lit.)
density   0.905 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NA 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
147.2 °F
Flash Point(C) 
64 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

242071 (R)-(+)-N,N-Dimethyl-1-phenylethylamine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?