• USA Home
  • 41389 - N,N-Dimethyl-L-phenylalanine

EMAIL THIS PAGE TO A FRIEND
41389 Sigma-Aldrich

N,N-Dimethyl-L-phenylalanine

puriss., ≥99.0% (sum of enantiomers, HPLC)

  • CAS Number 17469-89-5

  • Linear Formula C6H5CH2CH[N(CH3)2]CO2H

  • Molecular Weight 193.24

  •  Beilstein Registry Number 2966581

  •  MDL number MFCD00082539

  •  PubChem Substance ID 329756097

Purchase

Properties

InChI Key   HOGIQTACRLIOHC-JTQLQIEISA-N
grade   puriss.
assay   ≥99.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D +76±1°, c = 1.3% in H2O
mp   225-227 °C(lit.)
  225-228 °C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

273910 N,N-Dimethyl-L-phenylalanine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?