• USA Home
  • 413984 - Contract Lab Program AA/ICP Nitric acid reagent/matrix blank solution

EMAIL THIS PAGE TO A FRIEND
413984 Sigma-Aldrich

Contract Lab Program AA/ICP Nitric acid reagent/matrix blank solution

5% in ASTM type I water

Purchase

Properties

Related Categories AA/ICP Standard Solutions, AAS, Additional Standards, Analytical Standards, Analytical/Chromatography,
concentration   5% in ASTM type I water
application(s)   ICP/AAS: suitable
density   1.025 g/mL at 25 °C
format   multi-component solution

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2031 8/PG 2
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?