• USA Home
  • 41420 - 1,1-Dimethyl-4-phenylpiperazinium iodide

EMAIL THIS PAGE TO A FRIEND
41420 Sigma-Aldrich

1,1-Dimethyl-4-phenylpiperazinium iodide

≥99.0% (AT)

Synonym: DMPP

  • CAS Number 54-77-3

  • Empirical Formula (Hill Notation) C12H19IN2

  • Molecular Weight 318.20

  •  Beilstein Registry Number 3746109

  •  EC Number 200-213-0

  •  MDL number MFCD00011976

Purchase

Properties

Related Categories Chemical Synthesis, Heterocyclic Ammonium Salts, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   XFZJGFIKQCCLGK-UHFFFAOYSA-M
assay   ≥99.0% (AT)
mp   234-238 °C(lit.)
  236-238 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
TM3385000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?