EMAIL THIS PAGE TO A FRIEND
41430 Sigma-Aldrich

Dimethyl phosphite

purum, ≥98.0% (GC)

  • CAS Number 868-85-9

  • Linear Formula (CH3O)2P(O)H

  • Molecular Weight 110.05

  •  Beilstein Registry Number 1697490

  •  EC Number 212-783-8

  •  MDL number MFCD00044633

  •  PubChem Substance ID 329756104

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Organic Phosphates/Phosphites, Phosphorus Compounds More...
InChI Key   HZCDANOFLILNSA-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.402(lit.)
  n20/D 1.402
bp   170-171 °C(lit.)
density   1.2 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3278 6.1 / PGIII
WGK Germany 
1
RTECS 
SZ7710000
Flash Point(F) 
158 °F
Flash Point(C) 
70 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?