• USA Home
  • 41520 - 2,2-Dimethyl-1-propanol

EMAIL THIS PAGE TO A FRIEND
41520 Sigma-Aldrich

2,2-Dimethyl-1-propanol

purum, ≥97.0% (GC)

Synonym: tert-Butyl carbinol, Neopentyl alcohol

  • CAS Number 75-84-3

  • Linear Formula (CH3)3CCH2OH

  • Molecular Weight 88.15

  •  Beilstein Registry Number 1730984

  •  EC Number 200-907-3

  •  MDL number MFCD00004682

  •  PubChem Substance ID 24865983

Purchase

Properties

Related Categories Alcohols, Building Blocks, C2 to C6, Chemical Synthesis, Organic Building Blocks,
vapor pressure   16 mmHg ( 20 °C)
InChI Key   KPSSIOMAKSHJJG-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
bp   113-114 °C(lit.)
mp   52-56 °C(lit.)
  52-56 °C
density   0.818 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1325 - class 4.1 - PG 2 - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
1
Flash Point(F) 
82.4 °F
Flash Point(C) 
28 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?