CAS Number 1310-73-2
Linear Formula NaOH
Molecular Weight 40.00
MDL number MFCD00003548
PubChem Substance ID 24866001
Popular Documents: Specification Sheet (PDF)
| Related Categories | Acids & Bases, Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Essential Chemicals, |
| Quality Level | PREMIUM |
| vapor pressure | 3 mmHg ( 37 °C) |
| concentration | 50% in H2O |
| density | 1.515 g/mL at 25 °C |
1 L in poly bottle
25, 100, 500 mL in poly bottle
4, 18 L in poly drum
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?