• USA Home
  • 41600 - N,N-Dimethylsulfamoyl chloride

EMAIL THIS PAGE TO A FRIEND
41600 Sigma-Aldrich

N,N-Dimethylsulfamoyl chloride

purum, ≥98.5% (AT)

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonyl Halides, Sulfur Compounds More...
InChI Key   JFCHSQDLLFJHOA-UHFFFAOYSA-N
grade   purum
assay   ≥98.5% (AT)
refractive index   n20/D 1.452(lit.)
  n20/D 1.452
bp   114 °C/75 mmHg(lit.)
density   1.337 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3390 8(6.1) / PGI
WGK Germany 
3
RTECS 
WO7185500
Flash Point(F) 
201.2 °F
Flash Point(C) 
94 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?