• USA Home
  • 41670 - 2,5-Dimethylthiophene

EMAIL THIS PAGE TO A FRIEND
41670 Sigma-Aldrich

2,5-Dimethylthiophene

purum, ≥98.0% (GC)

Synonym: NSC 60689

  • CAS Number 638-02-8

  • Empirical Formula (Hill Notation) C6H8S

  • Molecular Weight 112.19

  •  Beilstein Registry Number 106450

  •  EC Number 211-313-9

  •  MDL number MFCD00005452

Purchase

Properties

Related Categories Building Blocks, C4 to C6, Chemical Synthesis, Heterocyclic Building Blocks, Thiophenes More...
InChI Key   GWQOOADXMVQEFT-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.512(lit.)
  n20/D 1.513
bp   134 °C/740 mmHg(lit.)
density   0.985 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1993 3/PG 3
WGK Germany 
3
Flash Point(F) 
75 °F
Flash Point(C) 
24 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

D188603 2,5-Dimethylthiophene, 98.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?