• USA Home
  • 41717 - N,N-Dimethyltriisopropylsilylamine

EMAIL THIS PAGE TO A FRIEND
41717 Sigma-Aldrich

N,N-Dimethyltriisopropylsilylamine

purum, ≥96% (GC)

Synonym: (Dimethylamino)triisopropylsilane

Purchase

Properties

InChI Key   RDGLSXXPVKFUJI-UHFFFAOYSA-N
grade   purum
assay   ≥96% (GC)
refractive index   n20/D 1.451(lit.)
bp   76-78 °C/8 mmHg(lit.)
mp   27-30 °C(lit.)
density   0.833 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
153 °F
Flash Point(C) 
67 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?