• USA Home
  • 41720 - N,N-Dimethyltrimethylsilylamine

EMAIL THIS PAGE TO A FRIEND
41720 Sigma-Aldrich

N,N-Dimethyltrimethylsilylamine

purum, ≥95.0% (GC)

Synonym: (Dimethylamino)trimethylsilane, N-(Trimethylsilyl)dimethylamine, TMSDMA

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Building Blocks, Chemical Synthesis, Derivatization Reagents,
InChI Key   KAHVZNKZQFSBFW-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.397(lit.)
bp   84 °C(lit.)
density   0.732 g/mL at 25 °C(lit.)

Description

Other Notes

Powerful silylating agent; For the silylation of silica

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2733PSN1 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
10.4 °F
Flash Point(C) 
-12 °C
Protocols & Articles

Articles

Silylating Reagents for GC-MS Compatible Derivatives of NSAIDs

Human and veterinary pharmaceuticals are a group of “emerging” contaminants,1, 2 some of which are produced in increasingly large volumes every year. The amounts produced are reaching quantities simi...
Luis-Alberto Martin
AnalytiX Volume 10 Article 2
Keywords: Anti-inflammatory agents, Derivatizations, Environmental, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Metabolites, Pesticides, Pharmaceutical, Silylations, Veterinary

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?